Learn More
Cpi International Peak Performance Inorganic Trace Metal Standard, Copper (Cu), 1000 ug/mL, 100 mL
SDP
Supplier:  Cpi International S44001000141
Encompass
Peak Performance Copper (Cu) single-element standard is provided at a concentration of 1,000 ppm and a volume of 100 mL.
Catalog No. NC9117715