Learn More
Electron Microscopy Sciences Single-End Seeker/Probe
SDP
Catalog No. 5028543
Encompass_Preferred
Single end stainless steel probe with a hexagonal handle which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip. Single end probe which comes with one fine point curved tip
Catalog No. 50-285-43 Supplier Electron Microscopy Sciences Supplier No. 7294501
Single end stainless steel probe with a hexagonal handle which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip. Single end probe which comes with one fine point curved tip