Learn More
Kuhner Shaker Inc SINGLE FEEDTUBE GLUCOSE 50 M
Catalog No. 502570089
Encompass
Single FeedTube, Glucose, 50 mL tube, 75 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0089 Supplier Kuhner Shaker Inc Supplier No. SMFT01001
Single FeedTube, Glucose, 50 mL tube, 75 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!