Learn More
EMBRIENT INC Single Flow Meter
SDP
Catalog No. NC9703850
Encompass
Used to control the gas flow of premixed gases. Calibrated for 1.0 to 25 liter per minute (LPM) (or 0.1-10 LPM) and is used for controlling flow rate of pre-mixed gases.
Catalog No. NC9703850 Supplier EMBRIENT INC Supplier No. SFM 3001
Used to control the gas flow of premixed gases. Calibrated for 1.0 to 25 liter per minute (LPM) (or 0.1-10 LPM) and is used for controlling flow rate of pre-mixed gases.