Learn More
Med Vet International Single Frosted Slides, One End, One Side, 144/Box, Case of 10 Boxes/
Catalog No. 503045131
Encompass_Preferred
Premiere brand slides are made of corrosion resistant glass with smoothly polished edges. Precleaned. 75x25mm approx. 1mm thick. Packed 144 per box. 10 boxes/case.
Catalog No. 50-304-5131 Supplier Med Vet International Supplier No. 9105CS
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Premiere brand slides are made of corrosion resistant glass with smoothly polished edges. Precleaned. 75x25mm approx. 1mm thick. Packed 144 per box. 10 boxes/case.