Learn More
Omnimed Inc Single Glove Box Holder/Dispenser
SDP
Catalog No. 501649553
Encompass_Preferred
Omnimed Glove Box Holders are built for heavy duty use. Constructed of durable stainless steel for an easy to care for finish that will withstand years of use and still look new. They are ideal for use in medical, lab and food prep functions or any area where cross contamination is an issue pr protective gloves are needed. These holders are simple to load and can be mounted vertically or horizontally on a variety of surfaces. Each unit accommodates 1 glove box . Dispensers measure 6 1/4H x 10W x 4D.
This item is not returnable. View return policy
Catalog No. 50-164-9553 Supplier Omnimed Inc Supplier No. 3053001
Omnimed Glove Box Holders are built for heavy duty use. Constructed of durable stainless steel for an easy to care for finish that will withstand years of use and still look new. They are ideal for use in medical, lab and food prep functions or any area where cross contamination is an issue pr protective gloves are needed. These holders are simple to load and can be mounted vertically or horizontally on a variety of surfaces. Each unit accommodates 1 glove box . Dispensers measure 6 1/4H x 10W x 4D.