Learn More
Electron Microscopy Sciences Veco Single Hole Grid A1500, Copper with 1500 µm hole
SDP
Catalog No. 50980455
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 1500 µm
Catalog No. 50-980-455 Supplier Electron Microscopy Sciences Supplier No. 1500CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 1500 µm