Learn More
Electron Microscopy Sciences Veco Single Hole Grid A2000, Copper with 2000 µm hole
SDP
Catalog No. 5028623
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 2000 µm
Catalog No. 50-286-23 Supplier Electron Microscopy Sciences Supplier No. 2000CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 2000 µm