Learn More
Electron Microscopy Sciences Veco Single Hole Grid A800, Copper with 800 µm hole
SDP
Catalog No. 50980134
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 800 µm
Catalog No. 50-980-134 Supplier Electron Microscopy Sciences Supplier No. 0800CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 800 µm