Learn More
Electron Microscopy Sciences Veco Single Hole Grid A800, Nickel with 800 µm hole
SDP
Catalog No. 50980135
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Hole: 800 µm
Catalog No. 50-980-135 Supplier Electron Microscopy Sciences Supplier No. 0800NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Hole: 800 µm