Learn More
HEPATOCHEM EvoluChem PhotoRedOx Box™ photoreactor
Catalog No. NC1463970
Encompass_Preferred
The EvoluChem PhotoRedOx Box™ is the photochemistry photoreactor choice for the chemist who seeks to standardize laboratory photochemical setups economically. A flexible design allows interchangeable LEDs from 365 nm to 808 nm and a wide variety of vials.
Catalog No. NC1463970 Supplier HEPATOCHEM Supplier No. HCK100601016
The EvoluChem PhotoRedOx Box™ is the photochemistry photoreactor choice for the chemist who seeks to standardize laboratory photochemical setups economically. A flexible design allows interchangeable LEDs from 365 nm to 808 nm and a wide variety of vials.