Learn More
Perkin Elmer US LLC Single Lip Seal Forming Tool for Standard 75 mL, 40 Bar Digestion Vessel Pressure Seals
Catalog No. 502111695
Encompass_Preferred
The Single Lip Seal Forming Tool is used to expand one 75 mL vessel seal at a time. This is shipped with all 75 mL vessel systems.
Catalog No. 50-211-1695 Supplier Perkin Elmer US LLC Supplier No. N3132015
Add to Cart
Add to Cart
The Single Lip Seal Forming Tool is used to expand one 75 mL vessel seal at a time. This is shipped with all 75 mL vessel systems.