Learn More
BINDER™ Single Measurement Temperature and Humidity Calibration Certificate
SDP
Catalog No. 502875666 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5666 KBF Series
50-287-5647 KBF PRO Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-287-5666 Supplier BINDER™ Supplier No. 80122437
Add to Cart
Add to Cart
Temperature and humidity measurement taken in center of chamber with certificate.
Calibration measurement taken from the center of the chamber at 25°C/60% RH, or at specified test values.
Specifications
For Use With (Equipment) KBF Series
Duration 1 Day
Product Type Calibration Certificate