Learn More
Ted Pella Inc Single Mount Storage Tube and Mailer, 10/pk
SDP
Catalog No. NC9506646
Encompass
Pin mounts with the standard 3.2mm (1/8") pin diameter are protected and held securely in the base (cap) of the tube. Can be used for pin mounts up to 18mm diameter. Pin mounts are sold separately. Inside dimension of clear tube: ø21mm (0.83"), max sample height on stub is 20mm (0.79"). Outside dimensions are ø23mm (0.9") with a height of 52mm (2.05"). Clear tube is made of polystyrene, cap is made of low density polyethylene (LDPE). Sold in packs of 10.
Catalog No. NC9506646 Supplier Ted Pella Inc Supplier No. 16630
Pin mounts with the standard 3.2mm (1/8") pin diameter are protected and held securely in the base (cap) of the tube. Can be used for pin mounts up to 18mm diameter. Pin mounts are sold separately. Inside dimension of clear tube: ø21mm (0.83"), max sample height on stub is 20mm (0.79"). Outside dimensions are ø23mm (0.9") with a height of 52mm (2.05"). Clear tube is made of polystyrene, cap is made of low density polyethylene (LDPE). Sold in packs of 10.