Learn More
Hach Company Hach Single Parameter Monitoring Panel (SPMP-TU) with SC4500 controller and TU5300sc turbidimeter
Catalog No. NC3739662 Shop All Hach Company Products
Encompass
The Hach Single Parameter Monitoring Panel (SPMP-TU) integrates a low-range laser turbidimeter and an advanced controller into a pre-engineered, ready-to-install enclosure for turbidity monitoring. It simplifies installation and operation for municipal water and process monitoring by providing a compact, durable, and electrically compatible solution.
  • Pre-engineered, ready-to-install panel for turbidity monitoring.
  • Integrated controller and low-range turbidimeter for streamlined measurements.
  • Compact enclosure with durable thermoplastic construction.
  • Operates on 100-240 VAC, 50/60 Hz.
  • Dimensions: 685.8 mm x 584.2 mm x 292.1 mm; weight: 13.8 kg.
  • Designed for municipal water and process control applications.
Catalog No. NC3739662 Supplier Hach Company Supplier No. SPMPTU
Add to Cart
Add to Cart
Request a Quote
The Hach Single Parameter Monitoring Panel (SPMP-TU) integrates a low-range laser turbidimeter and an advanced controller into a pre-engineered, ready-to-install enclosure for turbidity monitoring. It simplifies installation and operation for municipal water and process monitoring by providing a compact, durable, and electrically compatible solution.
  • Pre-engineered, ready-to-install panel for turbidity monitoring.
  • Integrated controller and low-range turbidimeter for streamlined measurements.
  • Compact enclosure with durable thermoplastic construction.
  • Operates on 100-240 VAC, 50/60 Hz.
  • Dimensions: 685.8 mm x 584.2 mm x 292.1 mm; weight: 13.8 kg.
  • Designed for municipal water and process control applications.