Learn More
Electron Microscopy Sciences Pipetter Single Place Sliding Guide, 7¼" x 4" x 2"
SDP
Catalog No. 50930343
Encompass_Preferred
Sliding guide provides easy microplate viewing for pipetting and liquid transfer. Easily align a Pipetter with microplate wells. Clearly marked with row letters for fast and easy orientation. Reduce eyestrain. Adjust the guide height to accommodate flat or v-bottom microplatea or PCR plates. Compatible with multichannel or single-channel pipetters.
Size: 7¼" x 4" x 2"
Catalog No. 50-930-343 Supplier Electron Microscopy Sciences Supplier No. 7049430
Sliding guide provides easy microplate viewing for pipetting and liquid transfer. Easily align a Pipetter with microplate wells. Clearly marked with row letters for fast and easy orientation. Reduce eyestrain. Adjust the guide height to accommodate flat or v-bottom microplatea or PCR plates. Compatible with multichannel or single-channel pipetters.
Size: 7¼" x 4" x 2"