Learn More
Perkin Elmer US LLC Single Position Thermostatted Cell Holder with Stirrer
Catalog No. 502112749
Encompass_Preferred
This holder is compatible with the LS 45/50/50B/55 spectrometers.
Catalog No. 50-211-2749 Supplier Perkin Elmer US LLC Supplier No. L2250141
This holder is compatible with the LS 45/50/50B/55 spectrometers.