Learn More
Mopec Single-pouch Specimen Bag
SDP
Catalog No. 22446164 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
6 x 6 in.
Includes:
Absorbing sheet
No Absorbing Sheet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
22-446-164 6 x 6 in. No Absorbing Sheet
22-446-128 6 x 6 in. Absorbing sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-164 Supplier Mopec Supplier No. FS081
Add to Cart
Add to Cart
Tamper-evident specimen bags designed to safely contain leaks from urine bottles.
  • Contains liquid in the event of a urine bottle leak, with absorbent sheets capable of absorbing the entire contents of a specimen bottle.
  • Each bag features a secure adhesive seal that cannot be reopened without damage
  • Available with optional absorbent sheets that gel on contact with liquid, effectively handling anything from small spills to full bottle contents.
Specifications
Dimensions (L x W) 6 x 6 in.
Product Type Speciman Bag
Includes No Absorbing Sheet
Quantity 200/Pk.