Learn More
VEOLIA WTS Single-point Calibration Standards Set1-mg/l
Catalog No. NC0140220
Encompass
Set includes: (1) Reagent Water Blank, (1) 1 mg C/L TOC Standard (as KHP), and (1) 1 mg C/L ICStandard (as Na2CO3).
Catalog No. NC0140220 Supplier VEOLIA WTS Supplier No. STD 9000101
Set includes: (1) Reagent Water Blank, (1) 1 mg C/L TOC Standard (as KHP), and (1) 1 mg C/L ICStandard (as Na2CO3).