Learn More
Thermco Products Inc SINGLE PT100 THERMOM -100/300C
Catalog No. NC0913005
Encompass_Preferred
Laboratory Temperature Standard, for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. Single & Dual Probe models. Single Pt100 Platinum Thermometer -100/300�C & �F x 0.1�C & �F
Catalog No. NC0913005 Supplier Thermco Products Inc Supplier No. ACCD370P
Laboratory Temperature Standard, for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. Single & Dual Probe models. Single Pt100 Platinum Thermometer -100/300�C & �F x 0.1�C & �F