Learn More
Innovative Research Inc Single Donor Human Red Blood Cells Unit CPDA1
Catalog No. 502036202
Encompass_Preferred
Single Donor Human Red Blood Cells from Innovative Research are separated from a standard unit (approximately 450ml starting volume) of whole blood. The whole blood is centrifuged into layers, and the red blood cells are manually extracted (no chemical separators are used). The resulting RBC volume is approximately 250ml. These cells are not washed or resuspended. This product is custom-collected per order.
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. If you would like any further testing or analysis performed prior to shipping, please let us know. 
Donor Specifications
Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to accommodate demographic requests from a variety of genders, race
Catalog No. 50-203-6202 Supplier Innovative Research Inc Supplier No. IWB3CPDA1UNIT
Single Donor Human Red Blood Cells from Innovative Research are separated from a standard unit (approximately 450ml starting volume) of whole blood. The whole blood is centrifuged into layers, and the red blood cells are manually extracted (no chemical separators are used). The resulting RBC volume is approximately 250ml. These cells are not washed or resuspended. This product is custom-collected per order.
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. If you would like any further testing or analysis performed prior to shipping, please let us know. 
Donor Specifications
Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to accommodate demographic requests from a variety of genders, race