Learn More
Innovative Research Inc Single Donor Human Red Blood Cells Washed 40ml
Catalog No. 502036200
Encompass_Preferred
Single Donor Human Red Blood Cells Washed from Innovative Research are separated from type O+ whole blood via centrifugation. They are washed with saline to remove the buffy coat and other debris, and are suspended at a 25% dilution of cells to Alsever's solution. 
Catalog No. 50-203-6200 Supplier Innovative Research Inc Supplier No. IWB3ALS40ML
Single Donor Human Red Blood Cells Washed from Innovative Research are separated from type O+ whole blood via centrifugation. They are washed with saline to remove the buffy coat and other debris, and are suspended at a 25% dilution of cells to Alsever's solution. 