Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Supplier:  Kuhner Shaker Inc SM108731
Encompass
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 1600 mL Thomson Optimum Growth flask
Catalog No. 50-257-0308