Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Supplier:  Kuhner Shaker Inc SM108730
Encompass
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 1000 mL Duran lab bottle or one Kuhner Shaker 1000 mL flask
Catalog No. 50-257-0309