Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Supplier: Kuhner Shaker Inc SM108729
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 1000 mL Erlenmeyer flask