Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Catalog No. 502570314
Encompass
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 250 mL Erlenmeyer flask
Catalog No. 50-257-0314 Supplier Kuhner Shaker Inc Supplier No. SM107324
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 250 mL Erlenmeyer flask