Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Supplier:  Kuhner Shaker Inc SM107167
Encompass
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 1000 mL Schott bottle or one 500 mL Erlenmeyer flask
Catalog No. 50-257-0316