Learn More
Kuhner Shaker Inc SINGLE REMOVABLE PIN-CLAMP FOR
Catalog No. 502570305
Encompass
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 3000 mL/5000 mL Fernbach flask
Catalog No. 50-257-0305 Supplier Kuhner Shaker Inc Supplier No. SM108734
Single removable pin-clamp for use with Kuhner Shaker Universal Trays. Holds one 3000 mL/5000 mL Fernbach flask