Learn More
Alkali Scientific Single Serve Charcoal Dextran
SDP
Catalog No. 503028604
Encompass_Preferred
CellProSingle ServeFBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single ServeFBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single ServeFBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProFetal Bovine Serum (FBS) offers charcoaldextran stripped fetal bovine serum (FBS) for researchers requiring low levels of certain steroid hormones Charcoaldextran stripping reduces the serum concentration of many hormones and certain growth factors such as estradiol cortisol corticosterone the B vitamins T3 T4 and prostaglandins Post-strip assay results are reported This serum is useful where these endogenous molecules may interfere with experimental work This serum may show some reduced growth-promotion ability for cells requi
Catalog No. 50-302-8604 Supplier Alkali Scientific Supplier No. FB125SCS
CellProSingle ServeFBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single ServeFBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single ServeFBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProFetal Bovine Serum (FBS) offers charcoaldextran stripped fetal bovine serum (FBS) for researchers requiring low levels of certain steroid hormones Charcoaldextran stripping reduces the serum concentration of many hormones and certain growth factors such as estradiol cortisol corticosterone the B vitamins T3 T4 and prostaglandins Post-strip assay results are reported This serum is useful where these endogenous molecules may interfere with experimental work This serum may show some reduced growth-promotion ability for cells requi