Learn More
Alkali Scientific Single Serve Dialyzed Fetal Bo
SDP
Supplier:  Alkali Scientific FB108S
Encompass_Preferred
CellProSingle Serve FBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single Serve FBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single Serve FBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProDialyzed FBS is manufactured from US sourced FBS using a proprietary process to reduce concentrations of small molecules such as cytokines nucleotides amino acids hormones small proteins and salts This creates a small molecule environment which provides greater definition and control Dialyzed FBS is ideal for applications where normal FBS small molecule concentrations may be disruptive It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines Tri
Catalog No. 50-302-7453