Learn More
Alkali Scientific Single ServeFetal Bovine Serum
SDP
Supplier:  Alkali Scientific FB60S
Encompass_Preferred
CellProSingle ServeFBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single ServeFBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single Serve FBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProU S Source Fetal Bovine Sera is collected entirely from USDA licensed abattoirs and processed within the United States Our Alkali Scientificserum is maintained at or below 40F from collection through sterile bottling to ensure superior performance It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines Our Premium US Origin FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages cancer and canc
Catalog No. 50-302-6916