Learn More
Grainger Single Shank Ball SS 1/16 in Rope dia.
Catalog No. 502768008
Encompass_Preferred
The LOOS Single Shank Ball 1/16 in is a single shank ball end mill with a 1/16 inch diameter. It is made of high speed steel and has a 1/4 inch shank diameter. The overall length is 1-1/2 inches. This end mill is designed for general purpose milling in most material including steel, stainless steel, cast iron, aluminum, copper, brass, plastics and other non-ferrous metals. It can be run at speeds up to 25,000 RPM and feed rates up to 80 IPM. The ball end geometry allows milling of 3D contoured shapes.
Catalog No. 50-276-8008 Supplier Grainger Supplier No. 16X624
The LOOS Single Shank Ball 1/16 in is a single shank ball end mill with a 1/16 inch diameter. It is made of high speed steel and has a 1/4 inch shank diameter. The overall length is 1-1/2 inches. This end mill is designed for general purpose milling in most material including steel, stainless steel, cast iron, aluminum, copper, brass, plastics and other non-ferrous metals. It can be run at speeds up to 25,000 RPM and feed rates up to 80 IPM. The ball end geometry allows milling of 3D contoured shapes.