Learn More
Grainger Single Shank Ball SS 1/8 in Rope dia.
Catalog No. 502767218
Encompass_Preferred
The LOOS Single Shank Ball 1/8 in Compatible is a single shank ball end mill with a 1/8 inch diameter. It is made of high speed steel and has a 1/8 inch shank diameter. The overall length is 1-1/2 inches. This end mill is designed for general purpose milling in most material types including steel, aluminum, wood, and plastic. It features two flutes and a ball end for milling contoured surfaces. The maximum RPM is 21,000.
Catalog No. 50-276-7218 Supplier Grainger Supplier No. 16X626
The LOOS Single Shank Ball 1/8 in Compatible is a single shank ball end mill with a 1/8 inch diameter. It is made of high speed steel and has a 1/8 inch shank diameter. The overall length is 1-1/2 inches. This end mill is designed for general purpose milling in most material types including steel, aluminum, wood, and plastic. It features two flutes and a ball end for milling contoured surfaces. The maximum RPM is 21,000.