Learn More
Grainger Single Shank Ball SS 3/16 in Rope dia.
Catalog No. 502767191
Encompass_Preferred
The LOOS Single Shank Ball 3/16 in is a ball end milling cutter with a single shank and a 3/16 inch diameter ball. It is made of high speed steel and has a 1/4 inch shank diameter. The overall length is 1-1/2 inches. This single flute end mill is designed for general purpose milling in most material types including steel, aluminum, wood, and plastic. It can be run at speeds up to 2100 RPM with feed rates up to 20 inches per minute. The ball end geometry allows milling of 3 dimensional contoured shapes.
Catalog No. 50-276-7191 Supplier Grainger Supplier No. 16X628
The LOOS Single Shank Ball 3/16 in is a ball end milling cutter with a single shank and a 3/16 inch diameter ball. It is made of high speed steel and has a 1/4 inch shank diameter. The overall length is 1-1/2 inches. This single flute end mill is designed for general purpose milling in most material types including steel, aluminum, wood, and plastic. It can be run at speeds up to 2100 RPM with feed rates up to 20 inches per minute. The ball end geometry allows milling of 3 dimensional contoured shapes.