Learn More
Grainger Single Shank Ball SS 3/32 in Rope dia.
Catalog No. 502767902
Encompass_Preferred
The LOOS Single Shank Ball 3/32 in is a ball end milling cutter with a 3/32 inch diameter ball end. It has a single shank design and is made of solid carbide for wear resistance and strength. The overall length is 1-1/2 inches and shank diameter is 1/8 inch. It has 2 flutes and can be used for light milling in steel, cast iron, aluminum and other non-ferrous metals. This ball end mill is ideal for machining three dimensional contours or difficult to reach surfaces.
Catalog No. 50-276-7902 Supplier Grainger Supplier No. 16X625
The LOOS Single Shank Ball 3/32 in is a ball end milling cutter with a 3/32 inch diameter ball end. It has a single shank design and is made of solid carbide for wear resistance and strength. The overall length is 1-1/2 inches and shank diameter is 1/8 inch. It has 2 flutes and can be used for light milling in steel, cast iron, aluminum and other non-ferrous metals. This ball end mill is ideal for machining three dimensional contours or difficult to reach surfaces.