Learn More
Grainger Single Shank Ball SS 5/16 in Rope dia.
Catalog No. 502768138
Encompass_Preferred
The Grainger 16X632 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a C-face mounting style. The motor dimensions are 9-1/4" long x 7-3/4" wide x 8-3/4" high and it weighs 38 lbs. The 16X632 electric motor is ideal for industrial applications requiring up to 3/4 horsepower.
Catalog No. 50-276-8138 Supplier Grainger Supplier No. 16X632
The Grainger 16X632 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a C-face mounting style. The motor dimensions are 9-1/4" long x 7-3/4" wide x 8-3/4" high and it weighs 38 lbs. The 16X632 electric motor is ideal for industrial applications requiring up to 3/4 horsepower.