Learn More
Grainger Single Shank Ball SS 5/32 in Rope dia.
Catalog No. 502767074
Encompass_Preferred
The Grainger Approved Single Shank Ball Pein Hammer is a 16 oz hammer with a fiberglass handle that absorbs shock and reduces user fatigue. It has a fully polished striking face to prevent marring. The hammer head and handle are securely fastened with a steel wedge for durability. This general purpose hammer is suitable for metal forming tasks like riveting, shaping and straightening metal. It measures 13-3/4 inches long overall.
Catalog No. 50-276-7074 Supplier Grainger Supplier No. 16X627
The Grainger Approved Single Shank Ball Pein Hammer is a 16 oz hammer with a fiberglass handle that absorbs shock and reduces user fatigue. It has a fully polished striking face to prevent marring. The hammer head and handle are securely fastened with a steel wedge for durability. This general purpose hammer is suitable for metal forming tasks like riveting, shaping and straightening metal. It measures 13-3/4 inches long overall.