Learn More
Grainger Single Shank Ball SS 7/32 in Rope dia.
Catalog No. 502767055
Encompass_Preferred
The Grainger Approved Single Shank Ball Pein Hammer is a 16 oz hammer with a fiberglass handle that absorbs shock and reduces user fatigue. It has a fully polished striking face to prevent chipping and mushrooming. The hammer head and handle are securely fastened with a steel shank. This is a general purpose hammer suitable for metal forming, riveting, shaping and many other applications. It meets ANSI standards for striking tools. The hammer has an overall length of 13 inches.
Catalog No. 50-276-7055 Supplier Grainger Supplier No. 16X629
The Grainger Approved Single Shank Ball Pein Hammer is a 16 oz hammer with a fiberglass handle that absorbs shock and reduces user fatigue. It has a fully polished striking face to prevent chipping and mushrooming. The hammer head and handle are securely fastened with a steel shank. This is a general purpose hammer suitable for metal forming, riveting, shaping and many other applications. It meets ANSI standards for striking tools. The hammer has an overall length of 13 inches.