Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 1000 µm hole, Molybdenum
SDP
Catalog No. 501926545
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 1000 um.
Material: Molybdenum
Catalog No. 50-192-6545 Supplier Electron Microscopy Sciences Supplier No. GA1000MO
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 1000 um.
Material: Molybdenum