Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid GS2/3x1, Mo, 606 x 1000 µm Slot
SDP
Catalog No. 5028650
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Molybdnum
Slot: 606 x 1000 µm
Catalog No. 50-286-50 Supplier Electron Microscopy Sciences Supplier No. G60610MO
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Molybdnum
Slot: 606 x 1000 µm