Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid GS2x0.5, Copper, 2000 x 500 µm Slot
SDP
Catalog No. 5028654
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 2000 x 500 µm
Catalog No. 50-286-54 Supplier Electron Microscopy Sciences Supplier No. G205CU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 2000 x 500 µm