Learn More
Electron Microscopy Sciences Veco Grid L2 x 1.5, Copper with 2000 x 1500 µm Oval Slot
SDP
Catalog No. 5028658
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 2000 x 1500 µm
Material: Copper
Catalog No. 50-286-58 Supplier Electron Microscopy Sciences Supplier No. 2015CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 2000 x 1500 µm
Material: Copper