Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid GS2/3x1, Nickel, 606 x 1000 µm Slot
SDP
Supplier:  Electron Microscopy Sciences G60610NI
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Nickel
Slot: 606 x 1000 µm
Catalog No. 50-286-57