Learn More
Electron Microscopy Sciences Veco Grid L0.2 x 1.5, Copper with 200 x 1500 µm Oval Slot
SDP
Catalog No. 50980118
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 200 x 1500 µm
Material: Copper
Catalog No. 50-980-118 Supplier Electron Microscopy Sciences Supplier No. 0215CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 200 x 1500 µm
Material: Copper