Learn More
Perkin Elmer US LLC Single Stage High Purity Brass Regulator for CO and H2
Supplier:  Perkin Elmer US LLC 00230253
Encompass_Preferred
PerkinElmer regulators are constructed of high purity brass barstock and have stainless steel diaphragms and metal-to-metal seals. They are suitable for use with high purity (>99.995% pure), noncorrosive gases. All regulators are supplied with standard CGA fittings. It is recommended that you consult your local gas supplier to determine the proper tank regulator required. All regulators have an outlet shutoff valve and terminate in a 14 inch NPT Swagelok fitting. They are designed for use with high purity carrier gas and feature a barstock body construction, stainless steel diaphragms, and metal-to-metal seals. Technical Specifications: Delivery Pressure Use: CO, H2 Delivery Pressure Range: 10-200 psig Cylinder Pressure Gauge: 0-400 psig Gauge: 0-4,000 psig
Catalog No. 50-211-7205