Learn More
Grainger 80/20 T-Slot Extruded Aluminum Profile, 1.5 in. x 1.5 in., 72 in. Length, 4-Slot, 1 ea
Catalog No. 502766463
Encompass_Preferred
This 1.50 in. x 1.50 in. square T-slot profile is a heavy-duty framing extrusion designed for clean room applications, machine guards, enclosures, and workstations. Constructed from 6063-T6 aluminum with an anodized finish, this profile features four open T-slots, one on each face, to provide versatile mounting options. The inclusion of align-a-grooves simplifies assembly by assisting in profile alignment.
  • Features four open T-slots
  • Includes align-a-grooves for simplified alignment
  • Compatible with all 15-series fasteners
  • Constructed of 6063-T6 aluminum with an anodized finish
  • Smooth surface finish resists dust buildup and simplifies cleaning
  • ESD safe design
Catalog No. 50-276-6463 Supplier Grainger Supplier No. 16U251
This 1.50 in. x 1.50 in. square T-slot profile is a heavy-duty framing extrusion designed for clean room applications, machine guards, enclosures, and workstations. Constructed from 6063-T6 aluminum with an anodized finish, this profile features four open T-slots, one on each face, to provide versatile mounting options. The inclusion of align-a-grooves simplifies assembly by assisting in profile alignment.
  • Features four open T-slots
  • Includes align-a-grooves for simplified alignment
  • Compatible with all 15-series fasteners
  • Constructed of 6063-T6 aluminum with an anodized finish
  • Smooth surface finish resists dust buildup and simplifies cleaning
  • ESD safe design