Learn More
Braintree Scientific Single Temp Controller, 38 degrees C, use with Cat#: GETP
SDP
Catalog No. 501954876
Encompass
Single Temp Power Supply - Provides a single constant plate temp. Use with Cat#: GETP, NOTE: Final plate temp is dependent on room temp. Choose from 30C, 34C, or 38C **Sold for research purposes only and not for human use**
Catalog No. 50-195-4876 Supplier Braintree Scientific Supplier No. GPP3
Single Temp Power Supply - Provides a single constant plate temp. Use with Cat#: GETP, NOTE: Final plate temp is dependent on room temp. Choose from 30C, 34C, or 38C **Sold for research purposes only and not for human use**