Learn More
Analytical Sales & Services Single UHPLC Assembly - 20cm
Catalog No. 5000804526
Encompass
Single UHPLC Assembly - 20cm Stainless Steel Tubing with 1 Finger Tight Fitting
Catalog No. 50-008-04526 Supplier Analytical Sales & Services Supplier No. 61220
Single UHPLC Assembly - 20cm Stainless Steel Tubing with 1 Finger Tight Fitting