Learn More
Foxx Life Sciences EZBio Single Use Closure Assembly, Disposable Aseptic Transfer Caps, 38-430 VersaCap with Vent and Dip Tube, EZTop Replacement for 500mL Foxx or Nalgene Bottles, Sterilized, 20/CS
Catalog No. 501178734
Encompass_Preferred
437-0016-FLS: EZBio, Single Use Cap Assembly, 38-430 VersaCap, PP, Vented w/ Tube for 500mL Bottle, 20/cs. Foxx Life Sciences EZbio Single Use Cap Assembly for 500mL Bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components and Gamma Irradiated in the United States. Assemblies take advantage of Foxx patented VersaCap technology which means there is no machining necessary to produce the product. Perfect for single use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate. Foxx also offers the ability to provide and produce custom components, assemblies, and variations in a timely manner. For more information please contact us at sales@foxxlifesciences.com or by phone at 1-603-890-3699.
Catalog No. 50-117-8734 Supplier Foxx Life Sciences Supplier No. 4300016FLS
437-0016-FLS: EZBio, Single Use Cap Assembly, 38-430 VersaCap, PP, Vented w/ Tube for 500mL Bottle, 20/cs. Foxx Life Sciences EZbio Single Use Cap Assembly for 500mL Bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components and Gamma Irradiated in the United States. Assemblies take advantage of Foxx patented VersaCap technology which means there is no machining necessary to produce the product. Perfect for single use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate. Foxx also offers the ability to provide and produce custom components, assemblies, and variations in a timely manner. For more information please contact us at sales@foxxlifesciences.com or by phone at 1-603-890-3699.