Learn More
Optek Danulat SINGLE USE CELL-SUC23-T1-1/4IN
Catalog No. NC3785776
Encompass
Single Use Cell - SUC23-T1, 1/4 inch Tube ID, Hose BarbModel: SUC23-TUBE1-0.250in-OPL000.0Hose barb fittings for 1/4 in. tubing inside diameter dimension • Fluid Path/Bore ID: Ø 0.177 in. (4.5 mm)- PPSU (polyphenylsulfone) body and fittings- 316L SS (1.4435) integrated conductivity electrode pins- Ø12 x 120 mm pH electrode holder, twist-lock- EPDM internal O-ring seals- USP <87> and <88> Class VI, FDA 21 CFR 177.2600- clean room manufactured (not sterile)- suitable for gamma irradiation, typical 25-40 kGy, tested to 55 kGyWetted Surface Finish: Ra < 0.8 µm (31.4 µinch)includes: Certificate 3.1 with material and conductivity datafor use with SUC Holder PRE L or R
Catalog No. NC3785776 Supplier Optek Danulat Supplier No. 40002302000000
Single Use Cell - SUC23-T1, 1/4 inch Tube ID, Hose BarbModel: SUC23-TUBE1-0.250in-OPL000.0Hose barb fittings for 1/4 in. tubing inside diameter dimension • Fluid Path/Bore ID: Ø 0.177 in. (4.5 mm)- PPSU (polyphenylsulfone) body and fittings- 316L SS (1.4435) integrated conductivity electrode pins- Ø12 x 120 mm pH electrode holder, twist-lock- EPDM internal O-ring seals- USP <87> and <88> Class VI, FDA 21 CFR 177.2600- clean room manufactured (not sterile)- suitable for gamma irradiation, typical 25-40 kGy, tested to 55 kGyWetted Surface Finish: Ra < 0.8 µm (31.4 µinch)includes: Certificate 3.1 with material and conductivity datafor use with SUC Holder PRE L or R